Learn More
Selleck Chemical LLC RGD (Arg-Gly-Asp) Peptides 10mg 99896-85-2
SDP
Supplier:  Selleck Chemical LLC S800810MG
Encompass_Preferred
RGD (Arg-Gly-Asp) Peptides is a cell adhesion motif which can mimic cell adhesion proteins and bind to integrins. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0969098