Learn More
Enzo Life Sciences RGD peptide (5 mg) CAS: 114681-65-1
SDP
Supplier:  Enzo Life Sciences BMLP7000005
Encompass_Preferred
Integrin ligand. Induces apoptosis in various cell types and cell lines at 1mM by a mechanism involving cellular uptake and possible direct stimulation of procaspase-3 autoprocessing. Does not induce apoptosis in the breast cancer line MCF-7, in which there is a functional deletion of the procaspase-3 gene. Contains the motif RGD (Arg-Gly-Asp) which is found in many integrin ligands. Inhibits cell adhesion of COS-7 cells to fibronectin 80-90% at 1mM. Alternative Name: GRGDNP. MW: 614.6. CAS: 114681-65-1. Formulation: Lyophilized. Purity: ≥97% (HPLC). Sequence Gly-Arg-Gly-Asp-Asn-Pro. Appearance: Solid. Solubility: Soluble in water (>50mg/ml). Long Term Storage: -20°C
Catalog No. NC0210557