Learn More
Advanced Biomatrix RGD Peptide Arg-Gly-Asp, 5mg, Sterile
Supplier:  Advanced Biomatrix 54625MG
Encompass_Preferred
PEPTITE-2000 is a synthetic peptide containing the RGD cell attachment sequence found in fibronectin, vitronectin and many other matrix and serum proteins. PEPTITE-2000 is supplied as 5 mg of sterile powder. This RGD peptide has been specifically engineered to spontaneously adhere to a variety of surfaces such as polystyrene (either tissue culture treated or non-treated), hollow fibers made of cellulose, glass, polycarbonate membranes and many other materials. This binding is mediated via a hydroscopic C-terminal sequence. The RGD motif is present at the N-terminal end of the peptide, allowing for optimal cell attachment via integrin receptors. PEPTITE-2000 can be substituted as the primary cell attachment component in a chemically defined animal component-free culture system.
Catalog No. 50-360-234