Learn More
Medchemexpress LLC HY-P1740A 1mg Medchemexpress, RGD peptide (GRGDNP) (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1740A1MG
Encompass_Preferred
Medchemexpress, HY-P1740A 1mg RGD peptide (GRGDNP) (TFA) CAS: RGD peptide (GRGDNP) (TFA) acts as an inhibitor of integrin-ligand interactions and plays an important role in cell adhesion, migration, growth, and differentiation [1] . RGD peptide (GRGDNP) (TFA) promote apoptosis through activation of conformation changes that enhance pro-caspase-3 activation and autoprocessing [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4243