Learn More
Apexbio Technology LLC RGD (Arg-Gly-Asp) Peptides,10mg CAS# 99896-85-2
SDP
Supplier:  Apexbio Technology LLC B370810
Encompass_Preferred
Synthetic peptides containing the arginine-glycine-aspartate (RGD) were extensively used as inhibitors of integrin-ligand interactions in studies of cell adhesion, migration, growth and differentiation, since the RGD motif is an integrin-recognition motif found in many ligands.In vitro: RGD peptide can induce apoptosis in the absence of signals and integrin-mediated cell clustering. Previous study demonstrates that RGD peptides promote apoptosis through activation of conformation changes enhancing pro-caspase-3 activation and autoprocessing [1]. In vivo: Anima study suggested that the RGD-4C-FITC-peptide bound to both endothelial and tumor cells in vivo and that peptide targeting should allow the delivery of therapeutic drugs to both endothelial and tumor cells [2]. Clinical trials: Currenlty no clinical data are available. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0071