Learn More
Cayman Chemical RGD PeptIdetrIfluoroacetAT 1mg
SDP
Catalog No. 502971735
Encompass_Preferred
A synthetic compound made up of the arginine-glycine-aspartate motif used to inhibit integrin-ligand interactions in studies related to cell adhesion, migration, growth, and differentiation; also can directly induce apoptosis by activating pro-caspase-3
Catalog No. 50-297-1735 Supplier Cayman Chemical Supplier No. 145011 MG
A synthetic compound made up of the arginine-glycine-aspartate motif used to inhibit integrin-ligand interactions in studies related to cell adhesion, migration, growth, and differentiation; also can directly induce apoptosis by activating pro-caspase-3