Learn More
Cayman Chemical RGD PeptIDtrIfluoROCTAte 25mg
SDP
Catalog No. 502973150
Encompass_Preferred
A synthetic compound made up of the arginine-glycine-aspartate motif used to inhibit integrin-ligand interactions in studies related to cell adhesion, migration, growth, and differentiation; also can directly induce apoptosis by activating pro-caspase-3
Catalog No. 50-297-3150 Supplier Cayman Chemical Supplier No. 1450125 MG
A synthetic compound made up of the arginine-glycine-aspartate motif used to inhibit integrin-ligand interactions in studies related to cell adhesion, migration, growth, and differentiation; also can directly induce apoptosis by activating pro-caspase-3