Learn More
Medchemexpress LLC HY-P0278A 10mg Medchemexpress, RGD Trifluoroacetate CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0278A10MG
Encompass_Preferred
Medchemexpress, HY-P0278A 10mg RGD Trifluoroacetate CAS: RGD Trifluoroacetate is a tripeptide that effectively triggers cell adhesion, addresses certain cell lines and elicits specific cell responses; RGD Trifluoroacetate binds to integrins . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4094