Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RGD (Trifluoroacetate) | 2378808-45-6 | 99.55% | 460.36 | 50 MG
SDP

Catalog No. 5000424977
Encompass_Preferred

RGD Trifluoroacetate is a tripeptide that effectively triggers cell adhesion, addresses certain cell lines, elicits specific cell responses, and binds to integrins. It is the most effective and most often employed peptide sequence for stimulated cell adhesion on synthetic surfaces. For research use only.

  • Effectively triggers cell adhesion
  • Addresses certain cell lines
  • Elicits specific cell responses
  • Binds to integrins, including α3β1, α5β1, α8β1, αIIbβ3, αvβ1, αvβ3, αvβ5, αvβ6, αvβ8, and to some extent α2β1 and α4β1
  • Suitable for laboratory centrifugation and storage

Catalog No. 50-004-24977 Supplier Medchemexpress LLC Supplier No. HYP0278A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RGD Trifluoroacetate is a tripeptide that effectively triggers cell adhesion, addresses certain cell lines, elicits specific cell responses, and binds to integrins. It is the most effective and most often employed peptide sequence for stimulated cell adhesion on synthetic surfaces. For research use only.

  • Effectively triggers cell adhesion
  • Addresses certain cell lines
  • Elicits specific cell responses
  • Binds to integrins, including α3β1, α5β1, α8β1, αIIbβ3, αvβ1, αvβ3, αvβ5, αvβ6, αvβ8, and to some extent α2β1 and α4β1
  • Suitable for laboratory centrifugation and storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.