Learn More
Aviva Systems Biology Corp INKA2 Antibody - middle region (ARP57803_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57803_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RGD1306526. It was validated on Western Blot. . Alternate names: RGD1306526ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-385