Learn More
Aviva Systems Biology Corp RGD1307041 Antibody - middle region (ARP57230_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57230_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RGD1307041. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: RGD1307041ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-283