Learn More
Aviva Systems Biology Corp RGD1310788 Antibody - C-terminal region (ARP35377_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35377_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RGD1310788. It was validated on Western Blot. . Alternate names: RGD1310788, ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-284-460