Learn More
Cayman Chemical RGDtargetd ProapoptoTCpep 10mg
SDP
Supplier:  Cayman Chemical 4213610 MG
Encompass_Preferred
A peptide; contains an integrin-binding cyclic region (CDCRGDCFC) and a region similar to a pro-apoptotic heptadimer sequence (klaklakklaklak), both of which are found in similar peptides
Catalog No. 50-294-5807