Learn More
Cayman Chemical RGD-targeted Proapoptotic Peptide, Purity: ≥95%, CAS: 2243207-09-0, 5 mg
SDP
Catalog No. 502945485
Encompass_Preferred
A peptide; contains an integrin-binding cyclic region (CDCRGDCFC) and a region similar to a pro-apoptotic heptadimer sequence (klaklakklaklak), both of which are found in similar peptides
Catalog No. 50-294-5485 Supplier Cayman Chemical Supplier No. 421365 MG
A peptide; contains an integrin-binding cyclic region (CDCRGDCFC) and a region similar to a pro-apoptotic heptadimer sequence (klaklakklaklak), both of which are found in similar peptides