Learn More
Abcam RGFP 966, HDAC3 inhibitor, 10MG
Catalog No. 502705499
Encompass_Preferred
MW 362.4 Da, Purity >99%. Potent, selective HDAC3 inhibitor (IC₅₀ = 80 nM). Induces DNA damage, impaired S phase progression and apoptosis in tumor cell lines. Shows central effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5499 Supplier Abcam Supplier No. AB14481910MG
MW 362.4 Da, Purity >99%. Potent, selective HDAC3 inhibitor (IC₅₀ = 80 nM). Induces DNA damage, impaired S phase progression and apoptosis in tumor cell lines. Shows central effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement