Learn More
Medchemexpress LLC HY-13909 10mg Medchemexpress, RGFP966 CAS:1357389-11-7 Purity:>98%
SDP
Catalog No. 502029037
Encompass_Preferred
Medchemexpress, HY-13909 10mg RGFP966 CAS:1357389-11-7 RGFP966 is a highly selective HDAC3 inhibitor with an IC50 of 80 nM and shows no inhibition to other HDACs at concentrations up to 15 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9037 Supplier Medchemexpress LLC Supplier No. HY1390910MG
Medchemexpress, HY-13909 10mg RGFP966 CAS:1357389-11-7 RGFP966 is a highly selective HDAC3 inhibitor with an IC50 of 80 nM and shows no inhibition to other HDACs at concentrations up to 15 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.