Learn More
Selleck Chemical LLC RGFP966 10mg 1396841-57-8
SDP
Supplier:  Selleck Chemical LLC S722910MG
Encompass_Preferred
RGFP966 is an HDAC3 inhibitor with IC50 of 0.08 ?M in cell-free assay, exhibits > 200-fold selectivity over other HDAC. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0574889