Learn More
Apexbio Technology LLC RGFP966, 200mg. Cas: 1357389-11-7;1396841-57-8 MFCD: MFCD26960954. Specific HDAC3 inhibitor
SDP
Supplier:  Apexbio Technology LLC A8803200
Encompass_Preferred
RGFP966 is a selective inhibitor of HDAC3 with IC50 value of 80 nM [1].HDAC3 (histone deacetylase 3) is an isoform of HDACs family and plays an important role in DNA transcriptional regulation. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5191