Learn More
Medchemexpress LLC HY-13909 5mg , RGFP966 CAS:1357389-11-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY139095MG
Encompass_Preferred
HY-13909 5mg RGFP966 CAS: 1357389-11-7 RGFP966 is a highly selective HDAC3 inhibitor with an IC50 of 80 nM and shows no inhibition to other HDACs at concentrations up to 15 μM. RGFP966 can penetrate the blood brain barrier (BBB). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9143