Learn More
TARGETMOL CHEMICALS INC RGFP966 E-ISOMER 5MG
SDP
Catalog No. 502579454
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. RGFP966 is an HDAC3 inhibitor with IC50 of 0.08 uM exhibits > 200-fold selectivity over other HDAC. purity: 99%
Catalog No. 50-257-9454 Supplier TARGETMOL CHEMICALS INC Supplier No. T62865MG
Also available in 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. RGFP966 is an HDAC3 inhibitor with IC50 of 0.08 uM exhibits > 200-fold selectivity over other HDAC. purity: 99%