Learn More
Aviva Systems Biology Corp RGL1 Antibody - C-terminal region (ARP64399_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64399_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RGL1. It was validated on Western Blot. . Alternate names: KIAA0959, RGLApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-255