Learn More
Aviva Systems Biology Corp RGL3 Antibody - middle region (ARP56251_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56251_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RGL3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RGL3. Alternate names: FLJ00153, FLJ32585, FLJ44275, MGC126805, MGC138163ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-286-130