Learn More
Medchemexpress LLC HY-139290 1mg , RGLS4326 CAS:2229964-07-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1392901MG
Encompass_Preferred
HY-139290 1mg RGLS4326 CAS: 2229964-07-0 RGLS4326 (RG4326) is a first-in-class, short oligonucleotide inhibitor of microRNA-17 (miR-17). RGLS4326 can be used for the research of autosomal dominant polycystic kidney disease (ADPKD). RGLS4326 inhibits miR-17 function in HeLa cells with an EC50 value of 28.3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1820