Learn More
Medchemexpress LLC RGLS4326 sodium | 99.7% | 3082.42 (free acid) | 1 MG
SDP
Catalog No. 5000349238
Encompass_Preferred
RGLS4326 sodium is a first-in-class, short oligonucleotide inhibitor of microRNA-17 (miR-17). It can be used for the research of autosomal dominant polycystic kidney disease (ADPKD).
  • First-in-class short oligonucleotide inhibitor
  • Inhibits microRNA-17 (miR-17)
  • Used for research of autosomal dominant polycystic kidney disease (ADPKD)
  • Inhibits miR-17 function in HeLa cells with an EC50 value of 28.3 nM
Catalog No. 50-003-49238 Supplier Medchemexpress LLC Supplier No. HY139290A1MG
RGLS4326 sodium is a first-in-class, short oligonucleotide inhibitor of microRNA-17 (miR-17). It can be used for the research of autosomal dominant polycystic kidney disease (ADPKD).
  • First-in-class short oligonucleotide inhibitor
  • Inhibits microRNA-17 (miR-17)
  • Used for research of autosomal dominant polycystic kidney disease (ADPKD)
  • Inhibits miR-17 function in HeLa cells with an EC50 value of 28.3 nM