Learn More
Aviva Systems Biology Corp RGMB Antibody - C-terminal region (ARP62193_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62193_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RGMB. It was validated on Western Blot. . Alternate names: DKFZp434P228, DRAGON, FLJ90406, MGC86970ApplicationWB. Species reactivity: Human
Catalog No. 50-287-606