Learn More
Enzo Life Sciences RGMC (human) monoclonal antibody (1D7) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2580100
Encompass_Preferred
Clone: 1D7. Isotype: IgG1. Immunogen: Recombinant human RGMC (aa 36-398) (ALX-522-104). Source: Purified from ascites. Species reactivity: Human. Applications: ELISA, Flow Cytometry, IP, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-2803