Learn More
Aviva Systems Biology Corp RGP1 Antibody - C-terminal region (ARP64931_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64931_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RGP1. It was validated on Western Blot. . Alternate names: KIAA0258ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-395