Learn More
Biomatik Corporation Rgpa Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11894
Encompass_Preferred
Rgpa Polyclonal Antibody is a Porphyromonas gingivalis Polyclonal Antibody to rgpA. The Rgpa Polyclonal Antibody has been validated for the following applications: ELISA. Host: Rabbit. Species: Porphyromonas gingivalis. Immunogen: Recombinant Porphyromonas Gingivalis Gingipain R1 Protein (228-720Aa). Applications: ELISA. Target Synonyms: rgpA. Accession Number: P28784. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5280