Learn More
Biomatik Corporation Rgpb Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11903
Encompass_Preferred
Rgpb Polyclonal Antibody is a Porphyromonas gingivalis Polyclonal Antibody to rgpB. The Rgpb Polyclonal Antibody has been validated for the following applications: ELISA. Host: Rabbit. Species: Porphyromonas gingivalis. Immunogen: Recombinant Porphyromonas Gingivalis Gingipain R2 Protein (230-473Aa). Applications: ELISA. Target Synonyms: rgpB. Accession Number: P95493. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5648