Learn More
Sino Biological RGS1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 15665T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human RGS1 (rh RGS1; Catalog#15665-H07E; NP_002913.3; Met1-Lys196). RGS1 specific IgG was purified by Human RGS1 affinity chromatography.
Catalog No. 50-224-7976