Learn More
Sino Biological RGS1 Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 15665RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human RGS1 (rh RGS1; Catalog#15665-H07E; NP_002913.3; Met1-Lys196). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-224-7532