Learn More
Aviva Systems Biology Corp Rgs10 Antibody - N-terminal region (ARP42856_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42856_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rgs10. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2310010N19Rik, MGC129414ApplicationFACS, WB. Species reactivity: Cow, Human, Mouse, Pig, Rat
Catalog No. 50-285-037