Learn More
Aviva Systems Biology Corp RGS13 Antibody - middle region (ARP34051_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34051_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RGS13. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human RGS13. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55231