Learn More
Aviva Systems Biology Corp RGS16 Antibody - C-terminal region (ARP34040_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34040_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RGS16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RGS16. Alternate names: RGS-R, A28-RGS14, A28-RGS14PApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-55225