Learn More
Aviva Systems Biology Corp Rgs18 Antibody - N-terminal region (ARP42853_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42853_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rgs18. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: MGC117531ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-285-036