Learn More
Aviva Systems Biology Corp RGS18 Antibody - middle region (ARP42805_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42805_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RGS18. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RGS18. Alternate names: RGS13ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Rat
Catalog No. 50-285-026