Learn More
Aviva Systems Biology Corp RGS19 Antibody - N-terminal region (ARP58934_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58934_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RGS19. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human RGS19. Alternate names: GAIP, RGSGAIPApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Rat
Catalog No. 50-286-533