Learn More
Aviva Systems Biology Corp Rgs2 Antibody - middle region (ARP42827_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42827_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against Rgs2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse Rgs2. Alternate names: AL592303.1, GOS8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-58345