Learn More
Aviva Systems Biology Corp Rgs2 Antibody - N-terminal region (ARP42826_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42826_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rgs2. It was validated on Western Blot. Rgs2,regulator of G-protein signaling 2,GOS8; Rgs2,regulator of G-protein signaling 2,regulator of G-protein signaling 2,OTTMUSP00000022856The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: GOS8ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-285-031