Learn More
Aviva Systems Biology Corp RGS20 Antibody - N-terminal region (ARP34054_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34054_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RGS20. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RGS20. Alternate names: RGSZ1, ZGAP1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55233