Learn More
Aviva Systems Biology Corp RGS20 Antibody - middle region (ARP34053_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34053_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RGS20. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human RGS20. Alternate names: RGSZ1, ZGAP1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55232