Learn More
Aviva Systems Biology Corp RGS3 Antibody - C-terminal region (AVARP09007_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP09007_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RGS3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RGS3. Alternate names: C2PA, FLJ20370, FLJ31516, FLJ90496, PDZ-RGS3, RGP3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62561