Learn More
Aviva Systems Biology Corp RGS6 Antibody - C-terminal region (ARP34043_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34043_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RGS6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human RGS6. Alternate names: GAPApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55227