Learn More
Aviva Systems Biology Corp RGS6 Antibody - N-terminal region (ARP34042_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34042_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RGS6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RGS6. Alternate names: DKFZp313G1241, FLJ43552, GAP, MGC142132ApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-55226