Learn More
Aviva Systems Biology Corp RGS8 Antibody - N-terminal region (ARP42803_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42803_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RGS8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RGS8. Alternate names: RP11-317P15.2, MGC119067, MGC119068, MGC119069ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58340