Learn More
Aviva Systems Biology Corp RGS9 Antibody - middle region (ARP30217_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30217_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RGS9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RGS9. Alternate names: MGC111763, MGC26458, PERRS, RGS9LApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-54144