Learn More
Aviva Systems Biology Corp RGS9BP Antibody - N-terminal region (ARP65375_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65375_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RGS9BP. It was validated on Western Blot. . Alternate names: FLJ45744, PERRS, R9AP, RGS9ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-488