Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Reagent 90, 0.016N Silver Nitrate in Acetone (4L)
SDP

Catalog No. NC0047660
Encompass_Preferred

Reagent 90, 0.016N Silver Nitrate in Acetone (4L)

Catalog No. NC0047660 Supplier Aqua Solutions Supplier No. DC876277
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Reagent 90, 0.016N Silver Nitrate in Acetone (4L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.