Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMarKa Chemical Company Reagent 91, Hydrolyzable Chloride Reactant 4l

Catalog No. 504400617
Encompass_Preferred

Reagent 91, Hydrolyzable Chloride Reactant-Bottle 4L Amber Safety

Catalog No. 50-440-0617 Supplier LaMarKa Chemical Company Supplier No. 00003947165282436
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Reagent 91, Hydrolyzable Chloride Reactant-Bottle 4L Amber Safety

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.