Learn More
Hach Company Aluminum Reagent Set, Eriochrome Cyanine R (ECR) Method, 50 mL
Catalog No. 501996005 Shop All Hach Company Products
Encompass
For determination of Aluminum by the Eriochrome Cyanine R (ECR) Method. Hach Method 8326. Sample Size: 50 mL Range: 0.002 - 0.220 mg/L. Reagent set contains ECR Reagent Powder Pillows, Hexamethylenetetramine Buffer Reagent Powder Pillows and ECR Masking Reagent Solution for approximately 100 tests.
Catalog No. 50-199-6005 Supplier Hach Company Supplier No. 2443400
For determination of Aluminum by the Eriochrome Cyanine R (ECR) Method. Hach Method 8326. Sample Size: 50 mL Range: 0.002 - 0.220 mg/L. Reagent set contains ECR Reagent Powder Pillows, Hexamethylenetetramine Buffer Reagent Powder Pillows and ECR Masking Reagent Solution for approximately 100 tests.